Assay Details
Target Gene Details
Entrez Gene ID: | 23794 |
Gene Name: | a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase-2) |
Gene Aliases: |
9530092O11Rik, ADAM-TS5, ADAMTS1, ADAMTS11, AI481094, ASMP-2 |
Location: |
Chr.16:85858157-85902374 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adamts5 | NM_011782.2 | NP_035912.2 | ||
AF140673.1 | AAD56356.1 | |||
AK044746.1 | BAC32062.1 | |||
AK082475.1 | BAC38503.1 | |||
BC138619.1 | ||||
BC138620.1 |
More Information
Set Membership: |
Intragenic Non-exonic |