Assay Details
Target Gene Details
Entrez Gene ID: | 84544 |
Gene Name: | CD96 antigen |
Gene Aliases: |
1700109I12Rik, Tactile |
Location: |
Chr.16:46035657-46120248 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd96 | NM_032465.2 | NP_115854.2 | ||
AK040539.1 | BAC30620.1 | |||
AK156470.1 | BAE33723.1 | |||
AY029156.1 | AAK31788.1 | |||
BC052865.1 | AAH52865.1 |
More Information
Set Membership: |
Intragenic Non-exonic |