Assay Details
Target Gene Details
Entrez Gene ID: | 16423 |
Gene Name: | CD47 antigen (Rh-related antigen, integrin-associated signal transducer) |
Gene Aliases: |
9130415E20Rik, AA407862, AI848868, AW108519, B430305P08Rik, IAP, Itgp |
Location: |
Chr.16:49855252-49912424 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd47 | NM_010581.3 | 1 | 89 | NP_034711.1 |
AB012693.1 | BAA25401.1 | |||
AK151922.1 | BAE30800.1 | |||
BC012667.1 | AAH12667.1 | |||
HQ585874.1 | ||||
Z25524.1 | 1 | 89 | CAA80978.1 |
More Information
Set Membership: |
Intragenic Exonic |