Assay Details
Target Gene Details
Entrez Gene ID: | 14167 |
Gene Name: | fibroblast growth factor 12 |
Gene Aliases: |
AV114868, B230343J05Rik, FGF-12, FHF-1, Fhf1 |
Location: |
Chr.16:28157783-28753243 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fgf12 | AF020739.1 | AAB71608.1 | ||
AK018774.1 | ||||
BB611389.1 | ||||
BC030485.1 | AAH30485.1 | |||
CX239635.1 |