Assay Details
Target Gene Details
Entrez Gene ID: | 13682 |
Gene Name: | eukaryotic translation initiation factor 4A2 |
Gene Aliases: |
4833432N07Rik, BM-010, Ddx2b, Eif4, eIF-4A-II, eIF4A-II |
Location: |
Chr.16:23107468-23114132 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif4a2 | NM_001123037.2 | NP_001116509.1 | ||
NM_001123038.2 | NP_001116510.1 | |||
NM_013506.3 | NP_038534.2 | |||
NR_110335.1 | ||||
AK076509.1 | BAC36372.1 | |||
AK088669.1 | BAC40492.1 | |||
AK132969.1 | BAE21449.1 | |||
AK165898.1 | ||||
BC094422.1 | AAH94422.1 | |||
CK623185.1 | ||||
X12507.1 | CAA31025.1 |