Assay Details
Target Gene Details
Entrez Gene ID: | 70511 |
Gene Name: | eukaryotic elongation factor 2 lysine methyltransferase |
Gene Aliases: |
5730409G15Rik, Fam86, Fam86a, eEF2-KMT |
Location: |
Chr.16:5244155-5255956 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eef2kmt | NM_027446.2 | 4 | 312 | NP_081722.1 |
AK017533.1 | 4 | 313 | BAB30795.3 | |
AK133590.1 | 4 | 313 | BAE21738.1 | |
AK163493.1 | 4 | 316 | BAE37370.1 | |
AK198006.1 | 4 | 264 | ||
BC051078.1 | 5 | 386 | AAH51078.1 |