Assay Details
Target Gene Details
Entrez Gene ID: | 67501 |
Gene Name: | coiled-coil domain containing 50 |
Gene Aliases: |
C3orf6 |
Location: |
Chr.16:27388765-27452218 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc50 | NM_001025615.3 | NP_001020786.1 | ||
NM_001289436.1 | NP_001276365.1 | |||
NM_026202.4 | NP_080478.2 | |||
AJ534985.1 | CAD59434.1 | |||
AK012190.1 | BAB28089.1 | |||
AK016827.1 | BAB30453.1 | |||
AK077557.1 | BAC36861.1 | |||
AK147820.1 | ||||
AK151672.1 | BAE30598.1 | |||
AK162434.1 | BAE36915.1 | |||
AK165214.1 | BAE38081.1 | |||
BC144895.1 | ||||
BC145933.1 | ||||
BC145935.1 | ||||
BY716501.1 | ||||
CJ045424.1 |