Assay Details
Target Gene Details
Entrez Gene ID: | 74147 |
Gene Name: | enoyl-Coenzyme A, hydratase/3-hydroxyacyl Coenzyme A dehydrogenase |
Gene Aliases: |
1300002P22Rik, HD, L-PBE, LBFP, LBP, MFP, MFP1, PBFE |
Location: |
Chr.16:21761285-21787834 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ehhadh | NM_023737.3 | NP_076226.2 | ||
AK004867.1 | BAB23628.1 | |||
BB841865.1 | ||||
BC016899.1 | AAH16899.1 |
More Information
Set Membership: |
Intragenic Non-exonic |