Assay Details
Target Gene Details
Entrez Gene ID: | 22122 |
Gene Name: | tissue specific transplantation antigen P35B |
Gene Aliases: |
AI256181, FX, Tstap35b |
Location: |
Chr.15:75924669-75929821 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tsta3 | NM_031201.1 | 2 | 158 | NP_112478.1 |
AK029632.1 | 2 | 182 | BAC26539.1 | |
BC089003.1 | 1 | 94 | AAH89003.1 | |
M30127.2 | 2 | 158 | AAA39673.2 |
More Information
Set Membership: |
Intragenic Exonic |