Assay Details
Target Gene Details
Entrez Gene ID: | 21819 |
Gene Name: | thyroglobulin |
Gene Aliases: |
Tgn, cog |
Location: |
Chr.15:66670756-66850721 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tg | NM_009375.2 | 20 | 4327 | NP_033401.2 |
AF076186.1 | 20 | 4305 | AAC32268.1 | |
AF076187.1 | 20 | 4305 | AAC32269.1 | |
BC111467.1 | 20 | 4320 | AAI11468.1 | |
U76389.1 | 20 | 4344 | AAB53204.1 |
More Information
Set Membership: |
Intragenic Exonic |