Assay Details
Target Gene Details
Entrez Gene ID: | 223701 |
Gene Name: | MKL (megakaryoblastic leukemia)/myocardin-like 1 |
Gene Aliases: |
AMKL, Bsac, Mal, Mrtf-A |
Location: |
Chr.15:81012281-81190757 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mkl1 | NM_001082536.1 | NP_001076005.1 | ||
NM_153049.3 | NP_694629.2 | |||
AF385582.1 | AAM94258.1 | |||
AF532597.1 | AAN33041.1 | |||
AK044188.1 | BAC31809.1 | |||
AK089416.1 | BAC40873.1 | |||
AK154561.1 | BAE32678.1 | |||
AK155821.1 | ||||
AK155937.1 | BAE33511.1 | |||
AK157985.1 | ||||
BC050941.1 | AAH50941.1 | |||
BY749697.1 |
More Information
Set Membership: |
Intragenic Non-exonic |