Assay Details
Target Gene Details
Entrez Gene ID: | 20491 |
Gene Name: | src-like adaptor |
Gene Aliases: |
Slap, Slap-1 |
Location: |
Chr.15:66780819-66831829 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sla | NM_001029841.4 | NP_001025012.1 | ||
NM_009192.3 | NP_033218.1 | |||
AJ131777.1 | CAB66139.1 | |||
AK036167.1 | BAC29328.1 | |||
AK037901.1 | BAC29896.1 | |||
AK041565.1 | BAC30988.1 | |||
AK137954.1 | BAE23522.1 | |||
AK163948.1 | ||||
BC032922.1 | AAH32922.1 | |||
CD578683.1 | ||||
CT010282.1 | CAJ18490.1 | |||
U29056.1 | AAA82756.1 |
Target Gene Details
Entrez Gene ID: | 21819 |
Gene Name: | thyroglobulin |
Gene Aliases: |
Tgn, cog |
Location: |
Chr.15:66670756-66850721 on Build GRCm38 |
Assay Gene Location: | Within Intron 41 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tg | NM_009375.2 | NP_033401.2 | ||
AF076186.1 | AAC32268.1 | |||
AF076187.1 | AAC32269.1 | |||
BC111467.1 | AAI11468.1 | |||
U76389.1 | AAB53204.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |