Assay Details
Target Gene Details
Entrez Gene ID: | 17749 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide K |
Gene Aliases: |
ABC10-alpha, MafY, Mt1a, RPABC4, RPB10alpha, RPB12, RPB7.0 |
Location: |
Chr.15:36174010-36177012 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polr2k | NM_001039368.2 | 1 | 100 | NP_001034457.1 |
NM_023127.3 | NP_075616.2 | |||
AK003023.1 | BAB22516.1 | |||
AK008136.1 | 1 | 57 | BAB25485.2 | |
AK008287.1 | 1 | 58 | BAB25578.2 | |
AK018914.1 | BAB31480.1 | |||
BY100585.1 | ||||
CR516049.1 | ||||
S63758.1 | AAB27565.2 |
More Information
Set Membership: |
Intragenic Exonic |