Assay Details
Target Gene Details
Entrez Gene ID: | 66218 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9 |
Gene Aliases: |
1190008J14Rik |
Location: |
Chr.15:58933810-58939489 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufb9 | NM_023172.3 | 4 | 579 | NP_075661.1 |
AJ300739.1 | 1 | 137 | CAC21577.1 | |
AK002254.1 | 4 | 580 | BAB21968.1 | |
AK003540.1 | 4 | 580 | BAB22846.1 | |
AK004522.1 | 4 | 580 | BAB23345.1 | |
BC031539.1 | 4 | 561 | AAH31539.1 |
More Information
Set Membership: |
Intragenic Exonic |