Assay Details
Target Gene Details
Entrez Gene ID: | 66218 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9 |
Gene Aliases: |
1190008J14Rik |
Location: |
Chr.15:58933810-58939489 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufb9 | NM_023172.3 | NP_075661.1 | ||
AK002254.1 | BAB21968.1 | |||
AK003540.1 | BAB22846.1 | |||
AK004522.1 | BAB23345.1 | |||
BC031539.1 | AAH31539.1 |
More Information
Set Membership: |
Intragenic Non-exonic |