Assay Details
Target Gene Details
Entrez Gene ID: | 69833 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide F |
Gene Aliases: |
1810060D16Rik |
Location: |
Chr.15:79141367-79151767 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polr2f | NM_027231.1 | NP_081507.1 | ||
AK007913.1 | BAB25345.1 | |||
AK165600.1 | BAE38285.1 | |||
AK169434.1 | BAE41175.1 | |||
BC024419.1 | AAH24419.1 |
More Information
Set Membership: |
Intragenic Non-exonic |