Assay Details
Target Gene Details
Entrez Gene ID: | 83797 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1 |
Gene Aliases: |
AA407987, Baf60a, D15Kz1 |
Location: |
Chr.15:99702287-99713995 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smarcd1 | NM_031842.2 | NP_114030.2 | ||
AK202807.1 | ||||
AK209291.1 | ||||
BC026783.1 | ||||
BC079839.1 | ||||
CV562378.1 | ||||
M25773.1 | AAA53377.1 | |||
U66620.1 | AAC52794.1 |
More Information
Set Membership: |
Intragenic Non-exonic |