Assay Details
Target Gene Details
Entrez Gene ID: | 105837 |
Gene Name: | Mdm2, transformed 3T3 cell double minute p53 binding protein |
Gene Aliases: |
AI429604, MDM2BP |
Location: |
Chr.15:55557408-55626423 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mtbp | NM_001168250.1 | 11 | 1299 | NP_001161722.1 |
NM_134092.3 | 11 | 1299 | NP_598853.3 | |
AJ278508.1 | 11 | 1220 | CAC07970.1 | |
AK080036.1 | 11 | 1113 | BAC37810.1 | |
AK088549.1 | 11 | 1113 | BAC40418.1 | |
AK088652.1 | 1 | 726 | BAC40479.1 | |
AK133855.1 | 11 | 1147 | BAE21889.1 | |
BC005500.1 | 4 | 296 | AAH05500.1 | |
BC052197.1 | 11 | 1299 | AAH52197.1 |