Assay Details
Target Gene Details
Entrez Gene ID: | 19387 |
Gene Name: | RAN GTPase activating protein 1 |
Gene Aliases: |
C79654, Fug1, mKIAA1835 |
Location: |
Chr.15:81704248-81729919 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rangap1 | NM_001146174.1 | 12 | 1565 | NP_001139646.1 |
NM_011241.4 | 13 | 1613 | NP_035371.4 | |
AK042930.1 | 12 | 1531 | ||
AK088768.1 | 12 | 1487 | BAC40559.1 | |
AK129452.1 | 12 | 1494 | BAC98262.1 | |
AK133905.1 | 12 | 1533 | BAE21919.1 | |
AK145459.1 | 11 | 1168 | BAE26449.1 | |
AK146670.1 | 13 | 1573 | BAE27347.1 | |
AK159009.1 | 12 | 1518 | BAE34767.1 | |
AK159525.1 | 12 | 1530 | BAE35154.1 | |
AK159930.1 | 12 | 1521 | ||
AK177694.1 | 2 | 96 | ||
AK179704.1 | 2 | 97 | ||
AK186071.1 | 2 | 97 | ||
BC014855.1 | 12 | 1521 | AAH14855.1 | |
BC052862.1 | 12 | 1488 | AAH52862.1 | |
BC066213.1 | 12 | 1529 | AAH66213.1 | |
BC071200.1 | 13 | 1729 | AAH71200.1 | |
U08110.1 | 13 | 1596 | AAA17681.1 |
More Information
Set Membership: |
Intragenic Exonic |