Assay Details
Target Gene Details
Entrez Gene ID: | 83456 |
Gene Name: | Moloney leukemia virus 10-like 1 |
Gene Aliases: |
CHAMP, Csm |
Location: |
Chr.15:88982891-89055152 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mov10l1 | NM_031260.2 | NP_112550.2 | ||
AF285587.1 | AAK31966.1 | |||
AF340211.1 | AAK77049.1 | |||
AK084786.1 | BAC39279.1 | |||
AY303754.1 | AAP60176.1 | |||
JF319145.1 |