Assay Details
Target Gene Details
Entrez Gene ID: | 71474 |
Gene Name: | protein phosphatase 6, regulatory subunit 2 |
Gene Aliases: |
1110033O10Rik, 8430411H09Rik, B230107H12Rik, Pp6r2, Saps2 |
Location: |
Chr.15:89211521-89287015 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp6r2 | NM_026813.1 | NP_081089.1 | ||
NM_027805.2 | NP_082081.1 | |||
AK018403.1 | ||||
AK083335.1 | BAC38872.1 | |||
AK084684.1 | BAC39251.1 | |||
BC024879.1 | AAH24879.1 |
More Information
Set Membership: |
Intragenic Non-exonic |