Assay Details
Target Gene Details
Entrez Gene ID: | 71474 |
Gene Name: | protein phosphatase 6, regulatory subunit 2 |
Gene Aliases: |
1110033O10Rik, 8430411H09Rik, B230107H12Rik, Pp6r2, Saps2 |
Location: |
Chr.15:89211521-89287015 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp6r2 | NM_026813.1 | 23 | 2960 | NP_081089.1 |
AK004079.1 | 4 | 377 | BAE43161.1 | |
AK173000.1 | 18 | 2143 | BAD32278.1 | |
BC024879.1 | 22 | 2763 | AAH24879.1 | |
BC112914.1 | 11 | 1119 |
More Information
Set Membership: |
Intragenic Exonic |