Assay Details
Target Gene Details
Entrez Gene ID: | 66513 |
Gene Name: | TGF-beta activated kinase 1/MAP3K7 binding protein 1 |
Gene Aliases: |
2310012M03Rik, Map3k7ip1, b2b449Clo |
Location: |
Chr.15:80133154-80161709 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tab1 | NM_025609.2 | NP_079885.2 | ||
AB088136.1 | BAC43729.1 | |||
BC027054.1 | AAH27054.1 | |||
BC041110.1 | AAH41110.1 | |||
BC054369.1 | AAH54369.1 |
More Information
Set Membership: |
Intragenic Non-exonic |