Assay Details
Target Gene Details
Entrez Gene ID: | 73062 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 16A |
Gene Aliases: |
2900084E10Rik, Mypt3, R75527 |
Location: |
Chr.15:76671620-76694919 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r16a | NM_033371.2 | NP_203535.1 | ||
AK150235.1 | ||||
AK154865.1 | BAE32888.1 | |||
AK171452.1 | BAE42461.1 | |||
AK184971.1 | ||||
AK220248.1 | BAD90173.1 | |||
AY010723.1 | AAG40949.1 | |||
BC033318.1 | AAH33318.1 | |||
BC057450.1 | AAH57450.1 | |||
BC096620.1 | AAH96620.1 |
More Information
Set Membership: |
Intragenic Non-exonic |