Assay Details
Target Gene Details
Entrez Gene ID: | 75475 |
Gene Name: | 5-oxoprolinase (ATP-hydrolysing) |
Gene Aliases: |
1700010G02Rik |
Location: |
Chr.15:76296603-76331101 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Oplah | NM_153122.2 | 19 | 2676 | NP_694762.1 |
AK164524.1 | 3 | 300 | BAE37822.1 | |
BC025120.1 | 6 | 774 | AAH25120.1 | |
BC034522.1 | 19 | 2676 | AAH34522.1 |
More Information
Set Membership: |
Intragenic Exonic |