Assay Details
Target Gene Details
Entrez Gene ID: | 107831 |
Gene Name: | adhesion G protein-coupled receptor B1 |
Gene Aliases: |
B830018M07Rik, Bai1, R75078, mKIAA4089 |
Location: |
Chr.15:74516196-74589465 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adgrb1 | NM_174991.3 | 18 | 3649 | NP_778156.2 |
AK046831.1 | 2 | 167 | BAC32888.1 | |
AK147456.1 | 18 | 3634 | BAE27923.1 | |
AK147459.1 | 18 | 3602 | BAE27926.1 | |
AK147494.1 | 18 | 3436 | BAE27949.1 | |
AK147600.1 | 18 | 3411 | ||
AK147607.1 | 18 | 3653 | BAE28021.1 | |
AK220481.1 | 18 | 3028 | BAD90505.1 | |
AY168408.1 | 18 | 3161 | AAN86966.1 | |
BC115835.1 |
More Information
Set Membership: |
Intragenic Exonic |