Assay Details
Target Gene Details
Entrez Gene ID: | 107831 |
Gene Name: | adhesion G protein-coupled receptor B1 |
Gene Aliases: |
B830018M07Rik, Bai1, R75078, mKIAA4089 |
Location: |
Chr.15:74516196-74589465 on Build GRCm38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adgrb1 | NM_174991.3 | NP_778156.2 | ||
AK147456.1 | BAE27923.1 | |||
AK147459.1 | BAE27926.1 | |||
AK147494.1 | BAE27949.1 | |||
AK147600.1 | ||||
AK147607.1 | BAE28021.1 | |||
AK220481.1 | BAD90505.1 | |||
AY168408.1 | AAN86966.1 | |||
BC115835.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |