Assay Details
Target Gene Details
Entrez Gene ID: | 54375 |
Gene Name: | antizyme inhibitor 1 |
Gene Aliases: |
1700085L02Rik, AZI, Oazi, Oazin |
Location: |
Chr.15:38487430-38519287 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Azin1 | NM_001102458.1 | NP_001095928.1 | ||
NM_001301688.1 | NP_001288617.1 | |||
NM_018745.5 | NP_061215.1 | |||
NR_125913.1 | ||||
AF032128.1 | AAB87464.1 | |||
AK049680.1 | BAC33870.1 | |||
AK088112.1 | BAC40151.1 | |||
AK088671.1 | BAC40494.1 | |||
AK133914.1 | ||||
BC019412.1 | AAH19412.1 | |||
BC043722.1 | AAH43722.1 | |||
CF738062.1 | ||||
CF749918.1 |
More Information
Set Membership: |
Intragenic Non-exonic |