Assay Details
Target Gene Details
Entrez Gene ID: | 68135 |
Gene Name: | eukaryotic translation initiation factor 3, subunit H |
Gene Aliases: |
1110008A16Rik, 40kD, 9430017H16Rik, EIF3-P40, EIF3-gamma, Eif3s3 |
Location: |
Chr.15:51786560-51865515 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif3h | NM_080635.1 | NP_542366.1 | ||
AK003560.1 | BAC25040.1 | |||
AK020033.1 | ||||
AK088436.1 | BAC40351.1 | |||
AK146200.1 | BAE26973.1 | |||
AK149081.1 | ||||
AK159113.1 | BAE34829.1 | |||
AK160335.1 | ||||
AK167651.1 | BAE39702.1 | |||
AK167815.1 | BAE39841.1 | |||
AK167844.1 | BAE39864.1 | |||
AK167928.1 | BAE39932.1 | |||
AK168108.1 | BAE40079.1 | |||
AK168123.1 | ||||
AK168259.1 | BAE40207.1 | |||
AK168274.1 | BAE40220.1 | |||
AK168553.1 | BAE40427.1 | |||
AK169416.1 | BAE41162.1 | |||
AK200513.1 | ||||
BC014755.1 | AAH14755.1 | |||
BC086915.1 | AAH86915.1 | |||
BC091728.1 | AAH91728.1 |
More Information
Set Membership: |
Intragenic Non-exonic |