Assay Details
Target Gene Details
Entrez Gene ID: | 239528 |
Gene Name: | argonaute RISC catalytic subunit 2 |
Gene Aliases: |
1110029L17Rik, 2310051F07Rik, AI225898, AL022874, AW546247, ENSMUSG00000072493, Eif2c2, Gerp95, Gm10365, mKIAA4215 |
Location: |
Chr.15:73101625-73184947 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ago2 | NM_153178.4 | NP_694818.3 | ||
AB081472.1 | BAC15767.1 | |||
AK220193.1 | BAD90378.1 | |||
BC024857.1 | AAH24857.2 | |||
BC064741.1 | AAH64741.1 | |||
BC096465.1 | AAH96465.1 | |||
BC128379.1 | ||||
BC129922.1 |
More Information
Set Membership: |
Intragenic Non-exonic |