Assay Details
Target Gene Details
Entrez Gene ID: | 109754 |
Gene Name: | cytochrome b5 reductase 3 |
Gene Aliases: |
0610016L08Rik, 2500002N19Rik, C85115, Dia-1, Dia1, WU:AL591952.1-001, WU:AL591952.1-002, WU:AL591952.1-003, WU:Cyb5r3 |
Location: |
Chr.15:83153495-83172208 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyb5r3 | NM_029787.2 | NP_084063.1 | ||
AF332059.1 | AAK56088.1 | |||
AF332060.1 | AAK56089.1 | |||
AK002640.1 | BAB22252.1 | |||
AK010858.1 | BAB27227.1 | |||
AK128917.1 | ||||
AK178355.1 | ||||
AK181262.1 | ||||
AK183532.1 | ||||
AK187049.1 | ||||
AK187743.1 | ||||
AK190014.1 | ||||
AK211950.1 | ||||
AK213922.1 | ||||
BC004760.1 | AAH04760.1 | |||
BC032013.1 | AAH32013.1 | |||
BC043074.1 | AAH43074.1 |
More Information
Set Membership: |
Intragenic Non-exonic |