Assay Details
Target Gene Details
Entrez Gene ID: | 18163 |
Gene Name: | catenin (cadherin associated protein), delta 2 |
Gene Aliases: |
Catnd2, Nprap, neurojugin |
Location: |
Chr.15:30172593-31029344 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ctnnd2 | NM_008729.2 | 11 | 2348 | NP_032755.2 |
AK140454.1 | BAE24395.1 | |||
BC138260.1 | 11 | 1893 | ||
BC138261.1 | 11 | 1893 | ||
BC145217.1 | 11 | 1890 | ||
U90331.1 | 11 | 2351 | AAB82409.1 |
More Information
Set Membership: |
Intragenic Exonic |