Assay Details
Target Gene Details
Entrez Gene ID: | 328572 |
Gene Name: | E1A binding protein p300 |
Gene Aliases: |
A430090G16, A730011L11, KAT3B, p300, p300 HAT |
Location: |
Chr.15:81586214-81652077 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 26 - Intron 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ep300 | NM_177821.6 | NP_808489.4 | ||
AK040384.1 | ||||
BC144976.1 | ||||
BC150681.1 | ||||
CD348567.1 |
More Information
Set Membership: |
Intragenic Non-exonic |