Assay Details
Target Gene Details
Entrez Gene ID: | 17444 |
Gene Name: | GRB2-related adaptor protein 2 |
Gene Aliases: |
GRAP-2, GRB2L, GRBLG, GRID, Gads, GrbX, Grf40, GrpL, Mona, P38 |
Location: |
Chr.15:80572422-80653047 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Grap2 | NM_001289442.1 | NP_001276371.1 | ||
NM_010815.3 | NP_034945.1 | |||
AF053405.1 | AAC98669.1 | |||
AF055465.1 | AAD08803.1 | |||
AF129477.1 | AAD41783.1 | |||
AF236118.1 | AAF60318.1 | |||
AJ011735.1 | CAA09756.1 | |||
AK156939.1 | BAE33906.1 | |||
BC052496.1 | AAH52496.1 |