Assay Details
Target Gene Details
Entrez Gene ID: | 66231 |
Gene Name: | THO complex 7 homolog (Drosophila) |
Gene Aliases: |
Nif3l1bp1 |
Location: |
Chr.14:13949012-13962042 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Thoc7 | NM_001285780.1 | NP_001272709.1 | ||
NM_025435.3 | NP_079711.1 | |||
AK005176.1 | BAB23863.1 | |||
AK007440.1 | BAB25040.1 | |||
AK010958.1 | BAB27291.1 | |||
AK013319.1 | ||||
AK045545.1 | ||||
AK143106.1 | 2 | 1576 | ||
AK147003.1 | BAE27600.1 | |||
BC042209.1 | AAH42209.1 | |||
BC054419.1 | AAH54419.1 | |||
BC116919.1 | ||||
BC116945.1 | ||||
CK329337.1 |