Assay Details
Target Gene Details
Entrez Gene ID: | 239337 |
Gene Name: | a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 12 |
Gene Aliases: |
ADAMTS-12, AI605170 |
Location: |
Chr.15:11064790-11346867 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adamts12 | NM_175501.2 | 16 | 2708 | NP_780710.2 |
AJ537452.1 | 16 | 2467 | CAD60967.1 | |
AK028459.1 | 7 | 963 | ||
AK048612.1 | 16 | 2671 | BAC33391.1 |
More Information
Set Membership: |
Intragenic Exonic |