Assay Details
Target Gene Details
Entrez Gene ID: | 76355 |
Gene Name: | TDP-glucose 4,6-dehydratase |
Gene Aliases: |
2610017J16Rik, 2610025M23Rik, AI648925 |
Location: |
Chr.14:118111911-118132765 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tgds | NM_029578.3 | NP_083854.3 | ||
AK011439.1 | ||||
AK011555.1 | BAB27693.1 | |||
AK154341.1 | BAE32527.1 | |||
AK165685.1 | ||||
AK165955.1 | BAE38483.1 | |||
BC021419.1 | AAH21419.1 | |||
BY710811.1 | ||||
CB953719.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |