Assay Details
Target Gene Details
Entrez Gene ID: | 110265 |
Gene Name: | methionine sulfoxide reductase A |
Gene Aliases: |
2310045J23Rik, 6530413P12Rik, MSR-A |
Location: |
Chr.14:64122621-64455903 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Msra | NM_001253712.1 | 5 | 680 | NP_001240641.1 |
NM_001253714.1 | 4 | 597 | NP_001240643.1 | |
NM_001253715.1 | 4 | 546 | NP_001240644.1 | |
NM_001253716.1 | 5 | 408 | NP_001240645.1 | |
NM_026322.4 | 10 | 1079 | NP_080598.2 | |
AK002356.1 | 5 | 555 | BAB22035.1 | |
AK009822.1 | 5 | 557 | BAB26522.1 | |
AK018338.1 | 5 | 402 | ||
AK049714.1 | 5 | 681 | BAC33889.1 | |
BC014738.1 | 4 | 505 | AAH14738.1 | |
BC089311.1 | 10 | 1079 | AAH89311.1 | |
BY282106.1 | ||||
CB953198.1 | 4 | 468 |
More Information
Set Membership: |
Intragenic Exonic |