Assay Details
Target Gene Details
Entrez Gene ID: | 56794 |
Gene Name: | 2-hydroxyacyl-CoA lyase 1 |
Gene Aliases: |
1600020H07Rik, Hpcl, Phyh2 |
Location: |
Chr.14:31607226-31641286 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hacl1 | NM_019975.3 | NP_064359.2 | ||
AJ132139.1 | CAB65550.1 | |||
AK005505.1 | BAB24085.1 | |||
AK041686.1 | BAC31032.1 | |||
AK050078.1 | BAC34059.1 | |||
BC021360.1 | AAH21360.1 |
More Information
Set Membership: |
Intragenic Non-exonic |