Assay Details
Target Gene Details
Entrez Gene ID: | 114874 |
Gene Name: | DDHD domain containing 1 |
Gene Aliases: |
Mir5131, PA-PLA1 |
Location: |
Chr.14:45593171-45658143 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddhd1 | NM_001039106.3 | NP_001034195.2 | ||
NM_001042719.2 | NP_001036184.1 | |||
NM_001284399.1 | NP_001271328.1 | |||
NM_176845.5 | NP_789815.3 | |||
AK014975.1 | ||||
AK048690.1 | ||||
AK129425.1 | BAC98235.1 | |||
AK155294.1 | ||||
AK170271.1 | BAE41676.1 | |||
BC018552.1 | AAH18552.1 | |||
BC043475.1 | AAH43475.1 | |||
CA315511.1 | ||||
DQ272481.1 | ABB76652.1 | |||
DQ272482.1 | ABB76653.1 |
More Information
Set Membership: |
Intragenic Non-exonic |