Assay Details
Target Gene Details
Entrez Gene ID: | 22070 |
Gene Name: | tumor protein, translationally-controlled 1 |
Gene Aliases: |
TCTP, Trt, p21, p23 |
Location: |
Chr.14:75845256-75848303 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tpt1 | NM_009429.3 | NP_033455.1 | ||
AK149695.1 | BAE29032.1 | |||
AK150300.1 | BAE29452.1 | |||
AK150500.1 | BAE29613.1 | |||
AK150858.1 | BAE29913.1 | |||
AK151059.1 | BAE30076.1 | |||
AK151266.1 | BAE30253.1 | |||
AK151375.1 | BAE30347.1 | |||
AK151894.1 | BAE30778.1 | |||
AK152690.1 | BAE31421.1 | |||
AK152808.1 | BAE31513.1 | |||
AK159516.1 | BAE35146.1 | |||
AK159565.1 | BAE35188.1 | |||
AK165736.1 | BAE38361.1 | |||
AK166584.1 | BAE38873.1 | |||
AK167413.1 | BAE39502.1 | |||
AK167936.1 | BAE39940.1 | |||
AK168151.1 | BAE40116.1 | |||
AK169730.1 | BAE41334.1 | |||
AK198939.1 | ||||
AK199313.1 | ||||
AK204488.1 | ||||
BC092381.1 | AAH92381.1 | |||
X06407.1 | CAA29697.1 |
More Information
Set Membership: |
Intragenic Non-exonic |