Assay Details
Target Gene Details
Entrez Gene ID: | 75901 |
Gene Name: | DCP1 decapping enzyme homolog A (S. cerevisiae) |
Gene Aliases: |
1110066A22Rik, 4930568L04Rik, AU019772, D14Ertd817e, Mitc1, SMIF |
Location: |
Chr.14:30479522-30527059 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dcp1a | NM_133761.3 | NP_598522.3 | ||
AJ344447.1 | CAC69875.1 | |||
AK047460.1 | ||||
AK171707.1 | BAE42622.1 | |||
DV651307.1 |
More Information
Set Membership: |
Intragenic Non-exonic |