Assay Details
Target Gene Details
Entrez Gene ID: | 75913 |
Gene Name: | RIKEN cDNA 4930579G18 gene |
Gene Aliases: |
- |
Location: |
Chr.14:54650511-54655100 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4930579G18Rik | NR_038053.1 | |||
AK016319.1 | BAB30192.1 |
Target Gene Details
Entrez Gene ID: | 56215 |
Gene Name: | apoptotic chromatin condensation inducer 1 |
Gene Aliases: |
2610036I19Rik, 2610510L13Rik, Acinus, Acn, C79325, acinusL, acinusS, mKIAA0670 |
Location: |
Chr.14:54642161-54686931 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acin1 | NM_001085472.2 | NP_001078941.2 | ||
NM_001085473.2 | NP_001078942.2 | |||
NM_001242605.1 | NP_001229534.1 | |||
NM_001242606.1 | NP_001229535.1 | |||
NM_019567.3 | NP_062513.3 | |||
NM_023190.3 | NP_075679.2 | |||
AF124725.1 | AAD56723.1 | |||
AF124729.1 | AAD56727.1 | |||
AF168782.1 | AAF89661.1 | |||
AK012099.1 | BAB28030.1 | |||
AK030449.1 | ||||
AK050467.1 | BAC34272.1 | |||
AK122342.1 | BAC65624.1 | |||
AV510393.1 | ||||
BC019127.1 | ||||
BC094217.1 | AAH94217.1 | |||
BY086848.1 | ||||
BY287113.1 | ||||
BY295579.1 | ||||
BY711288.1 | ||||
CJ161648.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |