Assay Details
Target Gene Details
Entrez Gene ID: | 20708 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 6b |
Gene Aliases: |
NK13, Spi12, ovalbumin |
Location: |
Chr.13:32956809-32979067 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Serpinb6b | NM_011454.1 | NP_035584.1 | ||
AK078753.1 | ||||
AK084785.1 | BAC39278.1 | |||
AK154048.1 | BAE32338.1 | |||
AK171741.1 | BAE42644.1 | |||
BC061050.1 | AAH61050.1 | |||
U96707.2 | AAB57819.2 |
More Information
Set Membership: |
Intragenic Non-exonic |