Assay Details
Target Gene Details
Entrez Gene ID: | 68750 |
Gene Name: | ras responsive element binding protein 1 |
Gene Aliases: |
1110037N09Rik, AA414966, B930013M22Rik |
Location: |
Chr.13:37778382-37952005 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rreb1 | NM_001039188.1 | NP_001034277.1 | ||
NM_001177868.1 | NP_001171339.1 | |||
NM_026830.2 | NP_081106.1 | |||
NR_033218.1 | 1 | 199 | ||
NR_033615.1 | ||||
AK047047.1 | ||||
AK047251.1 | ||||
AK147260.1 | 1 | 200 | BAE27803.1 | |
AK147653.1 | 1 | 200 | BAE28051.1 | |
AK154980.1 | BAE32969.1 | |||
AK170652.1 | ||||
AK171375.1 | BAE42417.1 | |||
BC080680.1 | AAH80680.1 | |||
BY202534.1 | ||||
BY205187.1 | ||||
BY206933.1 |