Assay Details
Target Gene Details
Entrez Gene ID: | 218030 |
Gene Name: | POU domain, class 6, transcription factor 2 |
Gene Aliases: |
D130006K24Rik, RPF-1 |
Location: |
Chr.13:18124960-18382044 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pou6f2 | NM_175006.2 | 6 | 1402 | NP_778171.2 |
AK042530.1 | 3 | 380 | BAC31283.1 | |
AK083772.1 | 6 | 1402 | BAC39017.1 | |
BC138151.1 | 6 | 1324 |