Assay Details
Target Gene Details
Entrez Gene ID: | 13162 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter, dopamine), member 3 |
Gene Aliases: |
DAT, Dat1 |
Location: |
Chr.13:73535528-73578672 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc6a3 | NM_010020.3 | NP_034150.1 | ||
AF109072.1 | AAD19643.1 | |||
AF109391.1 | AAF85795.1 | |||
AJ238309.1 | CAB51926.1 | |||
AK032629.1 | BAC27959.1 | |||
AK136885.1 | BAE23154.1 | |||
BC054119.1 | AAH54119.1 |
More Information
Set Membership: |
Intragenic Non-exonic |