Assay Details
Target Gene Details
Entrez Gene ID: | 13610 |
Gene Name: | sphingosine-1-phosphate receptor 3 |
Gene Aliases: |
AI132464, Edg3, Lpb3, S1p3 |
Location: |
Chr.13:51408618-51422797 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
S1pr3 | NM_010101.4 | 1 | 154 | NP_034231.1 |
AB028143.1 | BAA78207.1 | |||
AK028043.1 | BAC25715.1 | |||
AK029852.1 | BAC26645.1 | |||
AK030134.1 | BAC26800.1 | |||
AK047268.1 | BAC33008.1 | |||
AK078443.1 | BAC37277.1 | |||
AK081919.1 | BAC38373.1 | |||
AK083405.1 | ||||
AK084944.1 | BAC39316.1 | |||
AK085180.1 | BAC39383.1 | |||
AK087076.1 | ||||
AK087139.1 | ||||
BC064007.1 | AAH64007.1 | |||
BC068176.1 | AAH68176.1 |
More Information
Set Membership: |
Intragenic Exonic |