Assay Details
Target Gene Details
Entrez Gene ID: | 110596 |
Gene Name: | Rho guanine nucleotide exchange factor (GEF) 28 |
Gene Aliases: |
9230110L08Rik, AI323540, D13Bwg1089e, RIP2, Rgnef, RhoGEF, Rhoip2, p190RhoGEF |
Location: |
Chr.13:97898594-98206624 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arhgef28 | NM_012026.2 | 21 | 2590 | NP_036156.2 |
AK029167.1 | 15 | 1750 | ||
AK048676.1 | 13 | 1556 | ||
AK173325.1 | 21 | 2613 | BAD32603.1 | |
BC138030.1 | 21 | 2514 | ||
U73199.1 | 21 | 2590 | AAB18197.1 |