Assay Details
Target Gene Details
Entrez Gene ID: | 110596 |
Gene Name: | Rho guanine nucleotide exchange factor (GEF) 28 |
Gene Aliases: |
9230110L08Rik, AI323540, D13Bwg1089e, RIP2, Rgnef, RhoGEF, Rhoip2, p190RhoGEF |
Location: |
Chr.13:97898594-98206624 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arhgef28 | NM_012026.2 | NP_036156.2 | ||
AK029167.1 | ||||
AK048676.1 | ||||
AK173325.1 | BAD32603.1 | |||
BC022978.1 | ||||
BC075734.1 | AAH75734.1 | |||
BC138030.1 | ||||
U73199.1 | AAB18197.1 |